Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203405.1 | NP_001190334.1 | 1 - 2 | 756 | 100 | |
NM_001343580.1 | NP_001330589.1 | 1 - 2 | 709 | 100 | ||
NM_121905.3 | NP_197401.2 | 1 - 2 | 741 | 100 | ||
GenBank mRNA | AY136451.1 | - | 1 - 2 | 709 | 100 | |
BT002593.1 | - | 1 - 2 | 413 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |