Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPK16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121906.4 | NP_197402.1 | 8 - 9 | 1831 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |