Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IPT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343584.1 | NP_001318596.1 | 2 | 1521 | 102 | |
NM_001343585.1 | NP_001332154.1 | 2 | 1493 | 102 | ||
NM_121909.2 | NP_197405.1 | 1 | 1337 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |