Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AILP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036823.1 | NP_001031900.1 | 2 - 3 | 685 | 102 | |
NM_001343590.1 | NP_001330082.1 | 2 - 3 | 332 | 102 | ||
NM_121919.3 | NP_197415.1 | 2 - 3 | 569 | 102 | ||
GenBank mRNA | AY039946.1 | - | 2 - 3 | 565 | 102 | |
AY113899.1 | - | 2 - 3 | 440 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |