Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121927.3 | NP_197423.1 | 11 - 12 | 1249 | 82 | |
GenBank mRNA | AF370503.1 | - | 11 - 12 | 1248 | 82 | |
BT008884.1 | - | 11 - 12 | 1193 | 82 | ||
U72290.1 | - | 11 - 12 | 1206 | 82 | ||
X73367.2 | - | 1206 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |