Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121951.5 | NP_197447.2 | 2 - 3 | 705 | 140 | |
GenBank mRNA | AY074342.1 | - | 2 - 3 | 355 | 140 | |
AY117160.1 | - | 2 - 3 | 342 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |