Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343609.1 | NP_001331595.1 | 3 - 4 | 1827 | 84 | |
NM_121957.4 | NP_197453.1 | 3 - 4 | 1694 | 84 | ||
GenBank mRNA | AK228195.1 | - | 3 - 4 | 1628 | 84 | |
BT006457.1 | - | 2 - 3 | 1492 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |