Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128611.3 | NP_565703.1 | 2 - 3 | 1474 | 115 | |
GenBank mRNA | AF272747.1 | - | 2 - 3 | 1099 | 115 | |
AY136441.1 | - | 2 - 3 | 1416 | 115 | ||
BT000231.1 | - | 2 - 3 | 1001 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |