Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLE6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128664.2 | NP_565713.1 | 1 | 503 | 138 | |
GenBank mRNA | AY086725.1 | - | 1 | 477 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |