Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYCO ARATH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084517.1 | NP_001077986.1 | 4 - 5 | 796 | 74 | |
NM_001336323.1 | NP_001324768.1 | 2 - 3 | 717 | 74 | ||
NM_128673.5 | NP_565717.2 | 4 - 5 | 804 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |