Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPK24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128707.3 | NP_180708.1 | 3 - 4 | 939 | 86 | |
GenBank mRNA | DQ446585.1 | - | 3 - 4 | 661 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |