Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128714.4 | NP_180715.1 | 4 - 5 | 448 | 67 | |
GenBank mRNA | AY044330.1 | - | 4 - 5 | 300 | 67 | |
AY058187.1 | - | 4 - 5 | 403 | 67 | ||
AY086518.1 | - | 4 - 5 | 387 | 67 | ||
AY098982.1 | - | 4 - 5 | 300 | 67 | ||
U94495.1 | - | 4 - 5 | 325 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |