Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128714.4 | NP_180715.1 | 5 - 6 | 621 | 85 | |
GenBank mRNA | AY044330.1 | - | 5 - 6 | 473 | 85 | |
AY058187.1 | - | 5 - 6 | 576 | 85 | ||
AY086518.1 | - | 5 - 6 | 560 | 85 | ||
AY098982.1 | - | 5 - 6 | 473 | 85 | ||
U94495.1 | - | 5 - 6 | 498 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |