Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336351.1 | NP_001318329.1 | 17 - 18 | 2545 | 79 | |
NM_001336352.1 | NP_001323600.1 | 15 - 16 | 1832 | 79 | ||
NM_001336353.1 | NP_001323601.1 | 10 - 11 | 1058 | 79 | ||
NM_001336354.1 | NP_001323599.1 | 13 - 14 | 1623 | 79 | ||
NM_128748.3 | NP_180749.2 | 15 - 16 | 1832 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |