Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336351.1 | NP_001318329.1 | 4 - 5 | 987 | 80 | |
NM_001336352.1 | NP_001323600.1 | 2 - 3 | 274 | 80 | ||
NM_128748.3 | NP_180749.2 | 2 - 3 | 274 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |