Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336351.1 | NP_001318329.1 | 33 - 34 | 4657 | 126 | |
NM_001336352.1 | NP_001323600.1 | 31 - 32 | 3947 | 126 | ||
NM_001336353.1 | NP_001323601.1 | 26 - 27 | 3173 | 126 | ||
NM_001336354.1 | NP_001323599.1 | 29 - 30 | 3738 | 126 | ||
NM_128748.3 | NP_180749.2 | 31 - 32 | 3944 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |