Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336351.1 | NP_001318329.1 | 5 - 6 | 1132 | 65 | |
NM_001336352.1 | NP_001323600.1 | 3 - 4 | 419 | 65 | ||
NM_001336354.1 | NP_001323599.1 | 1 - 2 | 210 | 65 | ||
NM_128748.3 | NP_180749.2 | 3 - 4 | 419 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |