Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128796.3 | NP_180796.2 | 7 - 8 | 1329 | 83 | |
GenBank mRNA | EF988635.1 | - | 7 - 8 | 1329 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |