Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSLB04 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128813.2 | NP_180813.1 | 7 - 8 | 1496 | 79 | |
GenBank mRNA | KJ138880.1 | - | 7 - 8 | 1353 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |