Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHT1;5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128843.4 | NP_180842.1 | 1 - 2 | 412 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |