Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGLU33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128846.4 | NP_180845.2 | 1 - 2 | 382 | 75 | |
NM_201858.1 | NP_973587.1 | 1 - 2 | 382 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |