Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336434.1 | NP_001324212.1 | 37 - 38 | 5335 | 114 | |
NM_001336435.1 | NP_001324208.1 | 37 - 38 | 4881 | 114 | ||
NM_001336436.1 | NP_001324211.1 | 37 - 38 | 4902 | 114 | ||
NM_001336437.1 | NP_001324213.1 | 23 - 24 | 3392 | 114 | ||
NM_001336438.1 | NP_001324210.1 | 37 - 38 | 5163 | 114 | ||
NM_001336439.1 | NP_001324209.1 | 38 - 39 | 4857 | 114 | ||
NM_001336440.1 | NP_001324214.1 | 38 - 39 | 4857 | 114 | ||
NM_128883.3 | NP_180882.2 | 37 - 38 | 4920 | 114 | ||
GenBank mRNA | LC177513.1 | - | 36 - 37 | 4848 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |