Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BUBR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202739.2 | NP_001189668.1 | 4 - 5 | 698 | 154 | |
NM_128916.3 | NP_565767.1 | 4 - 5 | 698 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |