Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BUBR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202739.2 | NP_001189668.1 | 7 - 8 | 915 | 71 | |
NM_128916.3 | NP_565767.1 | 7 - 8 | 912 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |