Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336471.1 | NP_001324477.1 | 5 - 6 | 946 | 98 | |
NM_001336472.1 | NP_001324476.1 | 6 - 7 | 796 | 98 | ||
NM_128941.3 | NP_180938.2 | 6 - 7 | 772 | 98 | ||
GenBank mRNA | AY186582.1 | - | 6 - 7 | 698 | 98 | |
BT025874.1 | - | 6 - 7 | 698 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |