Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336475.1 | NP_001324363.1 | 1 - 2 | 501 | 91 | |
NM_128946.4 | NP_180942.1 | 2 - 3 | 745 | 91 | ||
GenBank mRNA | AF336917.1 | - | 2 - 3 | 380 | 91 | |
AF360313.1 | - | 2 - 3 | 478 | 91 | ||
AY056365.1 | - | 2 - 3 | 380 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |