Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336475.1 | NP_001324363.1 | 8 - 9 | 1380 | 83 | |
NM_128946.4 | NP_180942.1 | 9 - 10 | 1624 | 83 | ||
GenBank mRNA | AF336917.1 | - | 9 - 10 | 1259 | 83 | |
AF360313.1 | - | 9 - 10 | 1357 | 83 | ||
AY056365.1 | - | 9 - 10 | 1259 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |