Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: iqd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128950.5 | NP_180946.2 | 3 - 4 | 656 | 118 | |
GenBank mRNA | AK229389.1 | - | 3 - 4 | 323 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |