Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202741.2 | NP_001189670.2 | 2 - 3 | 397 | 75 | |
NM_001336484.1 | NP_001323468.1 | 2 - 3 | 397 | 75 | ||
NM_001336485.1 | NP_001323467.1 | 2 - 3 | 292 | 75 | ||
NM_128960.3 | NP_180956.2 | 2 - 3 | 387 | 75 | ||
NM_201866.2 | NP_973595.1 | 2 - 3 | 387 | 75 | ||
GenBank mRNA | BT002072.1 | - | 2 - 3 | 316 | 75 | |
BT015789.1 | - | 2 - 3 | 292 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |