Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIPK13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128969.2 | NP_180965.1 | 1 | 1487 | 88 | |
GenBank mRNA | AF295668.1 | - | 1 | 1274 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |