Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336496.1 | NP_001318348.1 | 1 - 2 | 2067 | 143 | |
NM_128973.3 | NP_180969.2 | 3 - 4 | 2466 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |