Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYP81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333499.1 | NP_001323219.1 | 3 - 4 | 776 | 86 | |
NM_104052.3 | NP_564597.1 | 4 - 5 | 629 | 86 | ||
GenBank mRNA | BT002917.1 | - | 4 - 5 | 598 | 86 | |
BT004352.1 | - | 4 - 5 | 409 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |