Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IAA18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104076.4 | NP_175607.1 | 1 - 2 | 867 | 130 | |
GenBank mRNA | AF367263.1 | - | 1 - 2 | 538 | 130 | |
AY056062.1 | - | 1 - 2 | 278 | 130 | ||
AY669797.1 | - | 1 - 2 | 278 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |