Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SK21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116006.4 | NP_567113.1 | 9 - 10 | 1309 | 71 | |
GenBank mRNA | AY084971.1 | - | 9 - 10 | 1285 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |