Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYSC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084855.2 | NP_001078324.1 | 7 - 8 | 887 | 94 | |
NM_001203223.1 | NP_001190152.1 | 7 - 8 | 1089 | 94 | ||
NM_116009.4 | NP_191703.1 | 8 - 9 | 1131 | 94 | ||
GenBank mRNA | AB024282.1 | - | 8 - 9 | 979 | 94 | |
AJ010505.1 | - | 8 - 9 | 1008 | 94 | ||
AK226606.1 | - | 8 - 9 | 1001 | 94 | ||
AY087208.1 | - | 8 - 9 | 1063 | 94 | ||
AY093094.1 | - | 8 - 9 | 1065 | 94 | ||
AY128782.1 | - | 8 - 9 | 925 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |