Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDG14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084857.2 | NP_001078326.1 | 5 - 6 | 1607 | 70 | |
NM_116039.4 | NP_191733.3 | 5 - 6 | 1563 | 70 | ||
GenBank mRNA | AB493660.1 | - | 5 - 6 | 1139 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |