Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084860.1 | NP_001078329.1 | 4 - 5 | 887 | 112 | |
NM_001203231.1 | NP_001190160.1 | 4 - 5 | 696 | 112 | ||
NM_001340137.1 | NP_001327149.1 | 3 - 4 | 821 | 112 | ||
NM_116074.2 | NP_191768.2 | 5 - 6 | 762 | 112 | ||
GenBank mRNA | BT033108.1 | - | 4 - 5 | 673 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |