Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATERDJ3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116125.3 | NP_191819.1 | 6 - 7 | 970 | 82 | |
GenBank mRNA | AY087640.1 | - | 6 - 7 | 831 | 82 | |
AY094429.1 | - | 6 - 7 | 800 | 82 | ||
AY125534.1 | - | 6 - 7 | 667 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |