Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116135.3 | NP_191829.1 | 4 - 5 | 3584 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |