Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116156.5 | NP_191850.2 | 14 - 15 | 1282 | 77 | |
GenBank mRNA | AB109893.1 | - | 14 - 15 | 1188 | 77 | |
AY090253.1 | - | 14 - 15 | 1229 | 77 | ||
AY125535.1 | - | 14 - 15 | 1188 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |