Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116156.5 | NP_191850.2 | 7 - 8 | 655 | 61 | |
GenBank mRNA | AB109893.1 | - | 7 - 8 | 561 | 61 | |
AY090253.1 | - | 7 - 8 | 602 | 61 | ||
AY125535.1 | - | 7 - 8 | 561 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |