Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSP41A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116179.5 | NP_191873.1 | 2 - 3 | 688 | 66 | |
GenBank mRNA | AF428269.1 | - | 2 - 3 | 547 | 66 | |
AY059887.1 | - | 2 - 3 | 610 | 66 | ||
AY128804.1 | - | 2 - 3 | 519 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |