Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIRO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116180.4 | NP_567139.1 | 6 - 7 | 1384 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |