Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116184.4 | NP_567141.1 | 6 - 7 | 761 | 75 | |
GenBank mRNA | AF372883.1 | - | 6 - 7 | 649 | 75 | |
AY056778.1 | - | 6 - 7 | 729 | 75 | ||
AY092982.1 | - | 6 - 7 | 667 | 75 | ||
BT002678.1 | - | 6 - 7 | 600 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |