Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116184.4 | NP_567141.1 | 7 - 8 | 811 | 101 | |
GenBank mRNA | AF372883.1 | - | 7 - 8 | 699 | 101 | |
AY056778.1 | - | 7 - 8 | 779 | 101 | ||
AY092982.1 | - | 7 - 8 | 717 | 101 | ||
BT002678.1 | - | 7 - 8 | 650 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |