Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340195.1 | NP_001319825.1 | 6 - 7 | 1086 | 99 | |
NM_116190.5 | NP_191884.1 | 6 - 7 | 976 | 99 | ||
NM_202753.2 | NP_974482.1 | 5 - 6 | 1372 | 99 | ||
GenBank mRNA | AF428402.1 | - | 6 - 7 | 923 | 99 | |
AK227627.1 | - | 6 - 7 | 925 | 99 | ||
BT010165.1 | - | 6 - 7 | 850 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |