Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116206.3 | NP_191900.1 | 1 - 2 | 668 | 59 | |
GenBank mRNA | AK316671.1 | - | 1 - 2 | 616 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |