Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116217.3 | NP_191911.1 | 12 - 13 | 1612 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |