Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YAB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116235.3 | NP_567154.1 | 1 - 2 | 603 | 76 | |
GenBank mRNA | AF136540.1 | - | 1 - 2 | 224 | 76 | |
AY037186.1 | - | 1 - 2 | 408 | 76 | ||
BT002662.1 | - | 1 - 2 | 116 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |