Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: iqd17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116308.4 | NP_567191.2 | 2 - 3 | 714 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |